Tisk…

5-Bromo-7-fluoro-2-(hydroxymethyl)benzofuran 95%

Dodavatel: Apollo Scientific
Varování

PC56923-1G PC56923-250MG
APOSPC56923-1GEA 13230 CZK
APOSPC56923-1G APOSPC56923-250MG
5-Bromo-7-fluoro-2-(hydroxymethyl)benzofuran 95%
5-Bromo-7-fluoro-2-(hydroxymethyl)benzofuran
Vzorec: C₉H₆BrFO₂
MW: 245,045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21233791
CAS číslo: 1498609-31-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR