Ethyl 5-bromo-7-fluoro-benzofuran-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
PC56917-1G
PC56917-250MG
APOSPC56917-1GEA
12730
CZK
APOSPC56917-1G
APOSPC56917-250MG
Ethyl 5-bromo-7-fluoro-benzofuran-2-carboxylate
Ethyl 5-bromo-7-fluoro-benzofuran-2-carboxylate
Vzorec:
C₁₁H₈BrFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD28100223 CAS číslo: 1404110-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...