3-Bromo-4-chloro-2-fluorobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC56902-1G
PC56902-250MG
PC56902-5G
PC56902-10G
APOSPC56902-1GEA
15710
CZK
APOSPC56902-1G
APOSPC56902-250MG
APOSPC56902-5G
APOSPC56902-10G
3-Bromo-4-chloro-2-fluorobenzaldehyde
3-Bromo-4-chloro-2-fluorobenzaldehyde
Vzorec:
C₇H₃BrClFO MW: 237,454 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28733866 CAS číslo: 1806838-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...