Tisk…

5-(Trifluoromethyl)nicotinaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(Trifluoromethyl)pyridine-3-carbaldehyde

PC56860-1G PC56860-5G
APOSPC56860-1GEA 10350 CZK
APOSPC56860-1G APOSPC56860-5G
5-(Trifluoromethyl)nicotinaldehyde
5-(Trifluoromethyl)nicotinaldehyde
Vzorec: C₇H₄F₃NO
MW: 175,108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696264
CAS číslo: 131747-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR