Tisk…

2-Chloro-4-(trifluoromethyl)benzylamine 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: [2-Chloro-4-(trifluoromethyl)phenyl]methanamine

PC56851-1G PC56851-250MG
APOSPC56851-1GEA 33400 CZK
APOSPC56851-1G APOSPC56851-250MG
2-Chloro-4-(trifluoromethyl)benzylamine 95%
2-Chloro-4-(trifluoromethyl)benzylamine
Vzorec: C₈H₇ClF₃N
MW: 209.596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06212820
CAS číslo: 581813-20-9
UN: 2735
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR