2-Bromo-5-iodobenzyl fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Bromo-2-(fluoromethyl)-4-iodobenzene
PC56850-1G
PC56850-250MG
APOSPC56850-1GEA
17360
CZK
APOSPC56850-1G
APOSPC56850-250MG
2-Bromo-5-iodobenzyl fluoride 95%
2-Bromo-5-iodobenzyl fluoride
Vzorec:
C₇H₅BrFI MW: 314.921 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31654714 CAS číslo: 2244085-57-0 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...