Tisk…

4-Bromo-2-(trifluoromethyl)phenylacetylene 95%

Dodavatel: Apollo Scientific
Varování

PC56843-1G PC56843-250MG
APOSPC56843-1GEA 2900 CZK
APOSPC56843-1G APOSPC56843-250MG
4-Bromo-2-(trifluoromethyl)phenylacetylene 95%
4-Bromo-2-(trifluoromethyl)phenylacetylene
Vzorec: C₉H₄BrF₃
MW: 249,027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31654734
CAS číslo: 2244085-78-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR