2-Bromo-5-(trifluoromethyl)phenylacetylene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC56841-1G
PC56841-250MG
APOSPC56841-1GEA
16340
CZK
APOSPC56841-1G
APOSPC56841-250MG
2-Bromo-5-(trifluoromethyl)phenylacetylene 95%
2-Bromo-5-(trifluoromethyl)phenylacetylene
Vzorec:
C₉H₄BrF₃ MW: 249.027 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30529953 CAS číslo: 1211523-00-0 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...