Tisk…

3-Bromo-4-fluorophenylacetylene 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-4-ethynyl-1-fluorobenzene

PC56838-1G PC56838-250MG
APOSPC56838-1GEA 16340 CZK
APOSPC56838-1G APOSPC56838-250MG
3-Bromo-4-fluorophenylacetylene 96%
3-Bromo-4-fluorophenylacetylene
Vzorec: C₈H₄BrF
Storage Temperature: Refrigerator
MDL Number: MFCD26395447
CAS číslo: 918487-39-5
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR