(2S)-2-Trifluoromethylpyrrolidine hydrochloride 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC56831-1G
PC56831-5G
APOSPC56831-1GEA
953
CZK
APOSPC56831-1G
APOSPC56831-5G
(2S)-2-Trifluoromethylpyrrolidine hydrochloride 97%
(2S)-2-Trifluoromethylpyrrolidine hydrochloride
Vzorec:
C₅H₈F₃N·ClH MW: 175,58 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22380026 CAS číslo: 1410903-57-9 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...