Tisk…

1-Bromo-4-(difluoromethyl)-2,5-difluorobenzene 95%

Dodavatel: Apollo Scientific
Varování

PC56825-1G PC56825-250MG
APOSPC56825-1GEA 18620 CZK
APOSPC56825-1G APOSPC56825-250MG
1-Bromo-4-(difluoromethyl)-2,5-difluorobenzene 95%
1-Bromo-4-(difluoromethyl)-2,5-difluorobenzene
Vzorec: C₇H₃BrF₄
Storage Temperature: Ambient
MDL Number: MFCD28737890
CAS číslo: 1628442-65-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR