2,3-Difluoro-5-methylbenzoic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC56815-1G
PC56815-250MG
APOSPC56815-1GEA
6850
CZK
APOSPC56815-1G
APOSPC56815-250MG
2,3-Difluoro-5-methylbenzoic acid 99%
2,3-Difluoro-5-methylbenzoic acid
Vzorec:
C₈H₆F₂O₂ MW: 172,129 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835284 CAS číslo: 1003709-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...