Tisk…

2,3-Difluoro-5-methylbenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC56811-1G PC56811-250MG PC56811-5G
APOSPC56811-1GEA 5290 CZK
APOSPC56811-1G APOSPC56811-250MG APOSPC56811-5G
2,3-Difluoro-5-methylbenzyl bromide
2,3-Difluoro-5-methylbenzyl bromide
Vzorec: C₈H₇BrF₂
Storage Temperature: Ambient
MDL Number: MFCD28790334
CAS číslo: 1806333-01-6
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR