2,3-Difluoro-5-methylbenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
PC56810-1G
PC56810-250MG
APOSPC56810-1GEA
16370
CZK
APOSPC56810-1G
APOSPC56810-250MG
2,3-Difluoro-5-methylbenzyl alcohol
2,3-Difluoro-5-methylbenzyl alcohol
Vzorec:
C₈H₈F₂O MW: 158,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16250146 CAS číslo: 1378816-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...