Tisk…

2,3,4-Trifluoro-6-(trifluoromethyl)benzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC56799-1G PC56799-250MG
APOSPC56799-1GEA 18620 CZK
APOSPC56799-1G APOSPC56799-250MG
2,3,4-Trifluoro-6-(trifluoromethyl)benzaldehyde 95%
2,3,4-Trifluoro-6-(trifluoromethyl)benzaldehyde
Vzorec: C₈H₂F₆O
MW: 228,091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31634834
CAS číslo: 2244084-38-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR