Tisk…

5-Bromo-3-fluoro-2-iodoanisole, tech

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-1-fluoro-2-iodo-3-methoxybenzene

PC56798-1G PC56798-5G
APOSPC56798-1GEA 1330 CZK
APOSPC56798-1G APOSPC56798-5G
5-Bromo-3-fluoro-2-iodoanisole, tech
5-Bromo-3-fluoro-2-iodoanisole
Vzorec: C₇H₅BrFIO
MW: 330,921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543687
CAS číslo: 1394291-47-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR