4-Bromobenzyl fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC56794-1G
PC56794-250MG
PC56794-5G
APOSPC56794-1GEA
6220
CZK
APOSPC56794-1G
APOSPC56794-250MG
APOSPC56794-5G
4-Bromobenzyl fluoride 95%
4-Bromobenzyl fluoride
Vzorec:
C₇H₆BrF MW: 189,025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20482167 CAS číslo: 459-49-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...