Tisk…

3-Bromobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC56792-1G PC56792-250MG
APOSPC56792-1GEA 35710 CZK
APOSPC56792-1G APOSPC56792-250MG
3-Bromobenzyl fluoride 95%
3-Bromobenzyl fluoride
Vzorec: C₇H₆BrF
Storage Temperature: Ambient
MDL Number: MFCD25963568
CAS číslo: 456-43-9
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR