Tisk…

2-Bromobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC56790-1G PC56790-250MG
APOSPC56790-1GEA 17360 CZK
APOSPC56790-1G APOSPC56790-250MG
2-Bromobenzyl fluoride 95%
2-Bromobenzyl fluoride
Vzorec: C₇H₆BrF
Storage Temperature: Ambient
MDL Number: MFCD26726839
CAS číslo: 446-47-9
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR