Tisk…

3-Nitrobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC56786-1G PC56786-250MG
APOSPC56786-1GEA 17360 CZK
APOSPC56786-1G APOSPC56786-250MG
3-Nitrobenzyl fluoride 95%
3-Nitrobenzyl fluoride
Vzorec: C₇H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD22053312
CAS číslo: 455-94-7
UN: 3265
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR