2-Chloro-4-(trifluoromethoxy)benzyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC56780-1G
PC56780-5G
APOSPC56780-1GEA
24870
CZK
APOSPC56780-1G
APOSPC56780-5G
2-Chloro-4-(trifluoromethoxy)benzyl bromide 95%
2-Chloro-4-(trifluoromethoxy)benzyl bromide
Vzorec:
C₈H₅BrClF₃O MW: 289.477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18393829 CAS číslo: 1261642-94-7 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...