Tisk…

tert-Butyl (R)-(1-fluoropropan-2-yl)carbamate 95%

Dodavatel: Apollo Scientific
Varování

PC56759-1G PC56759-250MG
APOSPC56759-1GEA 17360 CZK
APOSPC56759-1G APOSPC56759-250MG
tert-Butyl (R)-(1-fluoropropan-2-yl)carbamate 95%
tert-Butyl (R)-(1-fluoropropan-2-yl)carbamate
Vzorec: C₈H₁₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD30378923
CAS číslo: 2006287-03-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR