Tisk…

5-Bromo-2,3-difluoro-4-hydroxybenzaldehyde

Dodavatel: Apollo Scientific
Varování

PC56711-1G PC56711-5G
APOSPC56711-1GEA 39350 CZK
APOSPC56711-1G APOSPC56711-5G
5-Bromo-2,3-difluoro-4-hydroxybenzaldehyde
5-Bromo-2,3-difluoro-4-hydroxybenzaldehyde
Vzorec: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31618860
CAS číslo: 1895190-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR