Tisk…

1-Bromo-3,4-difluoro-2-iodobenzene 95%

Dodavatel: Apollo Scientific
Varování

PC56709-1G PC56709-250MG PC56709-5G
APOSPC56709-1GEA 15940 CZK
APOSPC56709-1G APOSPC56709-250MG APOSPC56709-5G
1-Bromo-3,4-difluoro-2-iodobenzene 95%
1-Bromo-3,4-difluoro-2-iodobenzene
Vzorec: C₆H₂BrF₂I
MW: 318,885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922588
CAS číslo: 1208077-27-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR