Tisk…

6-Bromo-3-fluoro-2-nitroaniline 95%

Dodavatel: Apollo Scientific
Varování

PC56703-1G PC56703-250MG
APOSPC56703-1GEA 14880 CZK
APOSPC56703-1G APOSPC56703-250MG
6-Bromo-3-fluoro-2-nitroaniline 95%
6-Bromo-3-fluoro-2-nitroaniline
Vzorec: C₆H₄BrFN₂O₂
MW: 235.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739830
CAS číslo: 1804841-33-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR