Tisk…

3-Bromo-2,5-difluorophenol 95+%

Dodavatel: Apollo Scientific
Varování

PC56338-1G PC56338-250MG
APOSPC56338-1GEA 20800 CZK
APOSPC56338-1G APOSPC56338-250MG
3-Bromo-2,5-difluorophenol 95+%
3-Bromo-2,5-difluorophenol
Vzorec: C₆H₃BrF₂O
MW: 208,988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28528419
CAS číslo: 1783357-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR