Tisk…

6-Bromo-3-fluoro-2-nitrophenol 95%

Dodavatel: Apollo Scientific
Varování

PC56337-1G
APOSPC56337-1GEA 4630 CZK
APOSPC56337-1G
6-Bromo-3-fluoro-2-nitrophenol 95%
6-Bromo-3-fluoro-2-nitrophenol
Vzorec: C₆H₃BrFNO₃
MW: 235,995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23701729
CAS číslo: 872363-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR