Tisk…

3-Chloro-2-(difluoromethoxy)aniline 95+%

Dodavatel: Apollo Scientific
Nebezpečí

PC56336-1G PC56336-250MG
APOSPC56336-1GEA 16340 CZK
APOSPC56336-1G APOSPC56336-250MG
3-Chloro-2-(difluoromethoxy)aniline 95+%
3-Chloro-2-(difluoromethoxy)aniline
Vzorec: C₇H₆ClF₂NO
MW: 193,578 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392559
CAS číslo: 288312-62-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR