6-Fluoro-2-methyl-3-nitrobenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC56285-1G
PC56285-250MG
PC56285-5G
APOSPC56285-1GEA
5590
CZK
APOSPC56285-1G
APOSPC56285-250MG
APOSPC56285-5G
6-Fluoro-2-methyl-3-nitrobenzoic acid 95%
6-Fluoro-2-methyl-3-nitrobenzoic acid
Vzorec:
C₈H₆FNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD28347722 CAS číslo: 1079992-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...