Tisk…

Methyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate 95%

Dodavatel: Apollo Scientific
Varování

PC56262-1G PC56262-250MG
APOSPC56262-1GEA 23970 CZK
APOSPC56262-1G APOSPC56262-250MG
Methyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate 95%
Methyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate
Vzorec: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31613607
CAS číslo: 2090828-24-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR