Tisk…

2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene 98%

Dodavatel: Apollo Scientific

PC56241-1G
APOSPC56241-1GEA 3810 CZK
APOSPC56241-1G
2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene 98%
2-Bromo-4-fluoro-1-(trifluoromethoxy)benzene
Vzorec: C₇H₃BrF₄O
MW: 258,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847187
CAS číslo: 866633-25-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR