Tisk…

4-Fluoro-3-phenoxybenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC56238-1G PC56238-250MG PC56238-5G
APOSPC56238-1GEA 18620 CZK
APOSPC56238-1G APOSPC56238-250MG APOSPC56238-5G
4-Fluoro-3-phenoxybenzoic acid 95%
4-Fluoro-3-phenoxybenzoic acid
Vzorec: C₁₃H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD03617287
CAS číslo: 77279-89-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR