Tisk…

5-Bromo-2-(trifluoromethoxy)anisole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-2-methoxy-1-(trifluoromethoxy)benzene

PC56210-1G PC56210-250MG
APOSPC56210-1GEA 9860 CZK
APOSPC56210-1G APOSPC56210-250MG
5-Bromo-2-(trifluoromethoxy)anisole
5-Bromo-2-(trifluoromethoxy)anisole
Vzorec: C₈H₆BrF₃O₂
MW: 271,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21333068
CAS číslo: 672948-65-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR