Tisk…

4-(1,1-Difluoroethyl)benzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC56188-1G PC56188-250MG
APOSPC56188-1GEA 25460 CZK
APOSPC56188-1G APOSPC56188-250MG
4-(1,1-Difluoroethyl)benzyl bromide
4-(1,1-Difluoroethyl)benzyl bromide
Vzorec: C₉H₉BrF₂
MW: 235,069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31568655
CAS číslo: 1654773-79-1
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR