1-(2,2,2-Trifluoroethyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
PC56183-1G
PC56183-5G
APOSPC56183-1GEA
5690
CZK
APOSPC56183-1G
APOSPC56183-5G
1-(2,2,2-Trifluoroethyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester
1-(2,2,2-Trifluoroethyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester
Vzorec:
C₁₃H₂₁BF₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18383322 CAS číslo: 1219931-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...