Tisk…

5-Bromo-6-fluoroquinoxaline

Dodavatel: Apollo Scientific
Nebezpečí

PC56171-1G PC56171-250MG
APOSPC56171-1GEA 20110 CZK
APOSPC56171-1G APOSPC56171-250MG
5-Bromo-6-fluoroquinoxaline
5-Bromo-6-fluoroquinoxaline
Vzorec: C₈H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD31568565
CAS číslo: 1893896-09-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR