Tisk…

2,3-Difluoro-4-iodobenzotrifluoride 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: Benzene, 2,3-difluoro-1-iodo-4-(trifluoromethyl)-

PC56170-1G PC56170-250MG PC56170-5G
APOSPC56170-1GEA 8010 CZK
APOSPC56170-1G APOSPC56170-250MG APOSPC56170-5G
2,3-Difluoro-4-iodobenzotrifluoride 95+%
2,3-Difluoro-4-iodobenzotrifluoride
Vzorec: C₇H₂F₅I
MW: 307,987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31568564
CAS číslo: 2227272-80-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR