Tisk…

Potassium [(4-methoxybenzyloxy)methyl]trifluoroborate

Dodavatel: Apollo Scientific
Varování

Synonyms: trifluoro-[(4-methoxyphenyl)methoxymethyl]boranuide, Potassium, Potassium trifluoro(((4-methoxybenzyl)oxy)methyl)borate

PC56166-5G PC56166-1G
APOSPC56166-5GEA 13460 CZK
APOSPC56166-5G APOSPC56166-1G
Potassium [(4-methoxybenzyloxy)methyl]trifluoroborate
Potassium [(4-methoxybenzyloxy)methyl]trifluoroborate
Vzorec: C₉H₁₁BF₃O₂·K
MW: 258,087 g/mol
Bod tání: 203…213 °C
Storage Temperature: Ambient
MDL Number: MFCD11505925
CAS číslo: 1027642-26-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR