Tisk…

2-Amino-4-(3-fluorophenyl)-1,3-thiazole

Dodavatel: Apollo Scientific
Varování

PC5616-1G PC5616-250MG
APOSPC5616-1GEA 3970 CZK
APOSPC5616-1G APOSPC5616-250MG
2-Amino-4-(3-fluorophenyl)-1,3-thiazole
2-Amino-4-(3-fluorophenyl)-1,3-thiazole
Vzorec: C₉H₇FN₂S
MW: 194,229 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094297
CAS číslo: 446065-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR