2-Bromo-3-fluoro-5-(hydroxymethyl)pyridine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(6-Bromo-5-fluoropyridine-3-yl)methanol
PC56155-1G
PC56155-250MG
PC56155-5G
APOSPC56155-1GEA
8270
CZK
APOSPC56155-1G
APOSPC56155-250MG
APOSPC56155-5G
2-Bromo-3-fluoro-5-(hydroxymethyl)pyridine 95+%
2-Bromo-3-fluoro-5-(hydroxymethyl)pyridine
Vzorec:
C₆H₅BrFNO MW: 206,012 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16606994 CAS číslo: 1227502-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...