2-Fluoro-4-iodophenol
Dodavatel: Apollo Scientific
|
Varování
|
PC56141-1G
PC56141-5G
PC56141-10G
APOSPC56141-1GEA
5000
CZK
APOSPC56141-1G
APOSPC56141-5G
APOSPC56141-10G
2-Fluoro-4-iodophenol
2-Fluoro-4-iodophenol
Vzorec:
C₆H₄FIO Storage Temperature: Freezer |
MDL Number:
MFCD11109769 CAS číslo: 2713-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...