Tisk…

2-Fluoro-5-hydrazinopyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: (6-Fluoropyridin-3-yl)hydrazine

PC56139-100MG PC56139-1G PC56139-250MG
APOSPC56139-100MGEA 5630 CZK
APOSPC56139-100MG APOSPC56139-1G APOSPC56139-250MG
2-Fluoro-5-hydrazinopyridine
2-Fluoro-5-hydrazinopyridine
Vzorec: C₅H₆FN₃
MW: 127.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848415
CAS číslo: 940958-93-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR