2-Fluoro-6-(trifluoromethoxy)benzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC56135-1G
PC56135-5G
APOSPC56135-1GEA
4570
CZK
APOSPC56135-1G
APOSPC56135-5G
2-Fluoro-6-(trifluoromethoxy)benzoic acid 97%
2-Fluoro-6-(trifluoromethoxy)benzoic acid
Vzorec:
C₈H₄F₄O₃ MW: 224.109 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22551466 CAS číslo: 1211530-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...