Tisk…

4-(Difluoromethoxy)-3-hydroxybenzoic acid 98%

Dodavatel: Apollo Scientific
Varování

PC56125-1G PC56125-250MG PC56125-5G
APOSPC56125-1GEA 3110 CZK
APOSPC56125-1G APOSPC56125-250MG APOSPC56125-5G
4-(Difluoromethoxy)-3-hydroxybenzoic acid 98%
4-(Difluoromethoxy)-3-hydroxybenzoic acid
Vzorec: C₈H₆F₂O₄
MW: 204,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24497448
CAS číslo: 913985-07-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR