2-Bromo-3,6-difluoronitrobenzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-1,4-difluoro-3-nitrobenzene
PC56123-1G
PC56123-250MG
APOSPC56123-1GEA
1450
CZK
APOSPC56123-1G
APOSPC56123-250MG
2-Bromo-3,6-difluoronitrobenzene 98%
2-Bromo-3,6-difluoronitrobenzene
Vzorec:
C₆H₂BrF₂NO₂ MW: 237,986 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28738282 CAS číslo: 1804402-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...