Methyl 3-(difluoromethoxy)-4-fluorobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC56117-1G
PC56117-250MG
APOSPC56117-1GEA
13460
CZK
APOSPC56117-1G
APOSPC56117-250MG
Methyl 3-(difluoromethoxy)-4-fluorobenzoate
Methyl 3-(difluoromethoxy)-4-fluorobenzoate
Vzorec:
C₉H₇F₃O₃ MW: 220,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399135 CAS číslo: 1261582-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...