3-Iodo-5-(trifluoromethyl)benzonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC56109-1G
PC56109-250MG
APOSPC56109-1GEA
20470
CZK
APOSPC56109-1G
APOSPC56109-250MG
3-Iodo-5-(trifluoromethyl)benzonitrile
3-Iodo-5-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₃F₃IN MW: 297,016 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12922961 CAS číslo: 691877-05-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...