Tisk…

2-(Difluoromethyl)-6-fluorobenzaldehyde 95+%

Dodavatel: Apollo Scientific
Varování

PC56105-1G PC56105-250MG
APOSPC56105-1GEA 17360 CZK
APOSPC56105-1G APOSPC56105-250MG
2-(Difluoromethyl)-6-fluorobenzaldehyde 95+%
2-(Difluoromethyl)-6-fluorobenzaldehyde
Vzorec: C₈H₅F₃O
MW: 174,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28519661
CAS číslo: 1785431-84-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR