Tisk…

2-(Bromomethyl)-6-fluoropyridine

Dodavatel: Apollo Scientific
Nebezpečí

PC56100-1G PC56100-250MG
APOSPC56100-1GEA 11540 CZK
APOSPC56100-1G APOSPC56100-250MG
2-(Bromomethyl)-6-fluoropyridine
2-(Bromomethyl)-6-fluoropyridine
Vzorec: C₆H₅BrFN
MW: 190,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953532
CAS číslo: 100202-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR