Tisk…

2-(Difluoro)oxirane 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3,3-Difluoro-1,2-epoxypropane

PC56095-1G PC56095-250MG
APOSPC56095-1GEA 41330 CZK
APOSPC56095-1G APOSPC56095-250MG
2-(Difluoro)oxirane 95%
2-(Difluoro)oxirane
Vzorec: C₃H₄F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD22414151
CAS číslo: 503-10-6
UN: 1992
ADR: 3+6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR